Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGCAGAAGGATCTCATGGCAGA[A/T]GGGATTCAGTGGCCAAGGAATGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602293 | ||||||||||||||||||||
Literature Links: |
CIB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CIB1 - calcium and integrin binding 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277764.1 | 296 | Intron | NP_001264693.1 | |||
NM_006384.3 | 296 | Intron | NP_006375.2 | |||
XM_006720375.1 | 296 | Intron | XP_006720438.1 |
GDPGP1 - GDP-D-glucose phosphorylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013657.2 | 296 | Missense Mutation | GAA,GAT | E,D 39 | NP_001013679.2 | |
NM_001322811.1 | 296 | Missense Mutation | GAA,GAT | E,D 39 | NP_001309740.1 |
TTLL13P - tubulin tyrosine ligase like 13, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |