Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCAGGGCCTCCATCTGGGGGTG[A/G]TGGGGCAGCCGCCAGGCCGGCGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604184 | ||||||||||||||||||||
Literature Links: |
IGDCC3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGDCC3 - immunoglobulin superfamily DCC subclass member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004884.3 | 2541 | Missense Mutation | CCA,TCA | P,S 784 | NP_004875.2 | |
XM_011522241.1 | 2541 | Missense Mutation | CCA,TCA | P,S 783 | XP_011520543.1 | |
XM_011522243.1 | 2541 | Missense Mutation | CCA,TCA | P,S 661 | XP_011520545.1 | |
XM_011522244.1 | 2541 | Missense Mutation | CCA,TCA | P,S 647 | XP_011520546.1 | |
XM_011522245.2 | 2541 | Intron | XP_011520547.1 |