Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCTCTCCCGTGTCCCTGGGGTC[C/T]GCTGGGGCAGCTGGCTCTTCACCGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C15orf59 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C15orf59 - chromosome 15 open reading frame 59 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039614.2 | 761 | Missense Mutation | CAG,CGG | Q,R 178 | NP_001034703.1 | |
NM_001303254.1 | 761 | Missense Mutation | CAG,CGG | Q,R 150 | NP_001290183.1 | |
XM_005254369.4 | 761 | Missense Mutation | CAG,CGG | Q,R 178 | XP_005254426.1 |