Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGCGCAGCGTGCGGAGGAACCT[A/G]AGCTACACGTGCCGCGCCAACCGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107773 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR1469 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR1469 - microRNA 1469 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR2F2 - nuclear receptor subfamily 2 group F member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145155.1 | 1557 | Intron | NP_001138627.1 | |||
NM_001145156.1 | 1557 | Intron | NP_001138628.1 | |||
NM_001145157.1 | 1557 | Intron | NP_001138629.1 | |||
NM_021005.3 | 1557 | Silent Mutation | CTA,CTG | L,L 111 | NP_066285.1 |
NR2F2-AS1 - NR2F2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |