Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGATGATGCCCCAGCTGCAGTTC[A/G]AAGATGCCTTTTGGGTGAGTGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606347 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSTPIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSTPIP1 - proline-serine-threonine phosphatase interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321135.1 | 472 | Missense Mutation | AAA,GAA | K,E 8 | NP_001308064.1 | |
NM_001321136.1 | 472 | UTR 5 | NP_001308065.1 | |||
NM_001321137.1 | 472 | Missense Mutation | AAA,GAA | K,E 73 | NP_001308066.1 | |
NM_003978.4 | 472 | Missense Mutation | AAA,GAA | K,E 8 | NP_003969.2 | |
XM_006720737.2 | 472 | UTR 5 | XP_006720800.1 | |||
XM_011522163.1 | 472 | Missense Mutation | AAA,GAA | K,E 8 | XP_011520465.1 | |
XM_011522164.1 | 472 | Intron | XP_011520466.1 | |||
XM_011522165.2 | 472 | Intron | XP_011520467.1 | |||
XM_011522166.1 | 472 | Missense Mutation | AAA,GAA | K,E 8 | XP_011520468.1 | |
XM_011522167.1 | 472 | Missense Mutation | AAA,GAA | K,E 8 | XP_011520469.1 | |
XM_011522168.2 | 472 | Missense Mutation | AAA,GAA | K,E 8 | XP_011520470.1 | |
XM_011522169.1 | 472 | Missense Mutation | AAA,GAA | K,E 8 | XP_011520471.1 |