Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGCCTCACCTACCCTTGGAAT[C/T]CTGGATATCATTGCACAGTACCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606934 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFAF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFAF1 - NADH:ubiquinone oxidoreductase complex assembly factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016013.3 | 892 | Silent Mutation | AGA,AGG | R,R 188 | NP_057097.2 | |
XM_006720555.2 | 892 | Silent Mutation | AGA,AGG | R,R 188 | XP_006720618.1 | |
XM_011521658.1 | 892 | Silent Mutation | AGA,AGG | R,R 188 | XP_011519960.1 | |
XM_011521659.2 | 892 | Silent Mutation | AGA,AGG | R,R 188 | XP_011519961.1 | |
XM_017022293.1 | 892 | Intron | XP_016877782.1 |