Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAAAAAAAAGAGGAAGCAGAAGT[A/G]AAACCAGGTGAGCCACTGGGGCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF3J PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF3J - eukaryotic translation initiation factor 3 subunit J | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284335.1 | 322 | Silent Mutation | GTA,GTG | V,V 65 | NP_001271264.1 | |
NM_001284336.1 | 322 | Intron | NP_001271265.1 | |||
NM_003758.3 | 322 | Silent Mutation | GTA,GTG | V,V 65 | NP_003749.2 |