Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGAGCGCACCGTCTTCTTCTTAG[A/T]CTGCACAGACAAGTAACTGAACAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142704 MIM: 603247 | ||||||||||||||||||||
Literature Links: |
HDC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HDC - histidine decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306146.1 | 1940 | Missense Mutation | ACT,TCT | T,S 544 | NP_001293075.1 | |
NM_002112.3 | 1940 | Missense Mutation | ACT,TCT | T,S 577 | NP_002103.2 | |
XM_017022094.1 | 1940 | Missense Mutation | ACT,TCT | T,S 612 | XP_016877583.1 | |
XM_017022095.1 | 1940 | Missense Mutation | ACT,TCT | T,S 579 | XP_016877584.1 | |
XM_017022096.1 | 1940 | Missense Mutation | ACT,TCT | T,S 536 | XP_016877585.1 | |
XM_017022097.1 | 1940 | Missense Mutation | ACT,TCT | T,S 533 | XP_016877586.1 | |
XM_017022098.1 | 1940 | Missense Mutation | ACT,TCT | T,S 468 | XP_016877587.1 | |
XM_017022099.1 | 1940 | Intron | XP_016877588.1 |
SLC27A2 - solute carrier family 27 member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |