Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACGGGAAGTGGAAAAACTCACACA[A/G]TGACTGGTTCTCCAGGGGAAGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605064 | ||||||||||||||||||||
Literature Links: |
KIF23 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF23 - kinesin family member 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281301.1 | 533 | Intron | NP_001268230.1 | |||
NM_004856.6 | 533 | Missense Mutation | ATG,GTG | M,V 122 | NP_004847.2 | |
NM_138555.3 | 533 | Missense Mutation | ATG,GTG | M,V 122 | NP_612565.1 | |
XM_005254796.2 | 533 | Missense Mutation | ATG,GTG | M,V 122 | XP_005254853.1 | |
XM_005254797.2 | 533 | Missense Mutation | ATG,GTG | M,V 122 | XP_005254854.1 | |
XM_005254798.2 | 533 | Missense Mutation | ATG,GTG | M,V 122 | XP_005254855.1 | |
XM_005254799.2 | 533 | Missense Mutation | ATG,GTG | M,V 122 | XP_005254856.1 | |
XM_011522238.1 | 533 | Intron | XP_011520540.1 | |||
XM_011522239.2 | 533 | Missense Mutation | ATG,GTG | M,V 1 | XP_011520541.1 | |
XM_011522240.1 | 533 | Intron | XP_011520542.1 | |||
XM_017022733.1 | 533 | Missense Mutation | ATG,GTG | M,V 122 | XP_016878222.1 |
LOC145694 - uncharacterized LOC145694 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |