Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGATGGCCCGCGCCCGCCTCGCCC[C/T]GGCCGGCATCCCTGCCGTCGCCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605123 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPINT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPINT1 - serine peptidase inhibitor, Kunitz type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032367.1 | 151 | Missense Mutation | CCG,CTG | P,L 14 | NP_001027539.1 | |
NM_003710.3 | 151 | Missense Mutation | CCG,CTG | P,L 14 | NP_003701.1 | |
NM_181642.2 | 151 | Missense Mutation | CCG,CTG | P,L 14 | NP_857593.1 | |
XM_006720657.1 | 151 | Missense Mutation | CCG,CTG | P,L 14 | XP_006720720.1 | |
XM_011521957.1 | 151 | Missense Mutation | CCG,CTG | P,L 14 | XP_011520259.1 | |
XM_011521958.1 | 151 | Missense Mutation | CCG,CTG | P,L 14 | XP_011520260.1 |