Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGAACACTGCATATCTTAATAAA[C/T]CCAGCCATTAGCATGATCAGAGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602192 | ||||||||||||||||||||
Literature Links: |
ADAM10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAM10 - ADAM metallopeptidase domain 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110.3 | 2526 | Silent Mutation | GGA,GGG | G,G 694 | NP_001101.1 | |
NM_001320570.1 | 2526 | Silent Mutation | GGA,GGG | G,G 663 | NP_001307499.1 | |
XM_017021861.1 | 2526 | Silent Mutation | GGA,GGG | G,G 620 | XP_016877350.1 |