Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTACCAGCAACGCTATGGGGCT[A/G]AGATAGTGAGGCGGTACAGGGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613880 MIM: 608429 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAHD1 - bromo adjacent homology domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHST14 - carbohydrate sulfotransferase 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130468.3 | 971 | Missense Mutation | AAG,GAG | K,E 240 | NP_569735.1 |