Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCTGGCCTTGGCTGCGCTCCGGA[A/G]TTCTTGGGTCGAAGAAGCAGGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610343 MIM: 608879 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2CD4A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C2CD4A - C2 calcium dependent domain containing 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207322.2 | 338 | Missense Mutation | AAT,AGT | N,S 66 | NP_997205.2 |
LOC101928907 - uncharacterized LOC101928907 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VPS13C - vacuolar protein sorting 13 homolog C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |