Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGGGGATCGGGCAGCTCTCTGC[C/T]CCCACGGCTCCAGCCTCAGCCCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607503 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DISP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DISP2 - dispatched RND transporter family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033510.2 | 2613 | Missense Mutation | CCC,TCC | P,S 119 | NP_277045.1 | |
XM_011522125.2 | 2613 | Missense Mutation | CCC,TCC | P,S 119 | XP_011520427.1 | |
XM_017022687.1 | 2613 | Missense Mutation | CCC,TCC | P,S 119 | XP_016878176.1 |
PHGR1 - proline, histidine and glycine rich 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |