Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCACCTGCACAGCAGACAGTGAG[A/G]TGCAGCGTTCGTAGATCTCCTTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604447 MIM: 610022 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNB5 - G protein subunit beta 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100129973 - uncharacterized LOC100129973 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYO5C - myosin VC | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018728.3 | 5138 | Missense Mutation | ACC,ATC | T,I 1659 | NP_061198.2 | |
XM_011521781.2 | 5138 | Missense Mutation | ACC,ATC | T,I 1631 | XP_011520083.1 | |
XM_017022408.1 | 5138 | Missense Mutation | ACC,ATC | T,I 1614 | XP_016877897.1 | |
XM_017022409.1 | 5138 | Missense Mutation | ACC,ATC | T,I 1372 | XP_016877898.1 | |
XM_017022410.1 | 5138 | Missense Mutation | ACC,ATC | T,I 1195 | XP_016877899.1 |