Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCAGCTTCTTCTCCATCTCCCCG[C/T]AGGACTGCCAGTTTGCTTTGGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190030 MIM: 600988 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FES PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FES - FES proto-oncogene, tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAN2A2 - mannosidase alpha class 2A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320977.1 | 508 | Nonsense Mutation | CAG,TAG | Q,* 114 | NP_001307906.1 | |
NM_006122.3 | 508 | Nonsense Mutation | CAG,TAG | Q,* 114 | NP_006113.2 | |
XM_005254910.3 | 508 | Nonsense Mutation | CAG,TAG | Q,* 114 | XP_005254967.1 | |
XM_011521565.2 | 508 | Intron | XP_011519867.1 | |||
XM_011521566.2 | 508 | Intron | XP_011519868.1 | |||
XM_017022184.1 | 508 | Nonsense Mutation | CAG,TAG | Q,* 114 | XP_016877673.1 | |
XM_017022185.1 | 508 | Nonsense Mutation | CAG,TAG | Q,* 114 | XP_016877674.1 |