Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAATGGATGGGGTGTCTTCAGA[A/G]GCTAATGAAGAAAAGTAAGTTCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609173 | ||||||||||||||||||||
Literature Links: |
KNL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KNL1 - kinetochore scaffold 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144508.4 | 182 | Silent Mutation | GAA,GAG | E,E 7 | NP_653091.3 | |
NM_170589.4 | 182 | Silent Mutation | GAA,GAG | E,E 7 | NP_733468.3 | |
XM_017022432.1 | 182 | UTR 5 | XP_016877921.1 |