Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTTGGGGCTCTGGTGACCAGGTTC[A/G]CAGCTCTGCTCCACGTTCTCCTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607961 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6881 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6881 - microRNA 6881 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEMA7A - semaphorin 7A (John Milton Hagen blood group) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146029.2 | 1698 | Silent Mutation | TGC,TGT | C,C 573 | NP_001139501.1 | |
NM_001146030.2 | 1698 | Silent Mutation | TGC,TGT | C,C 422 | NP_001139502.1 | |
NM_003612.4 | 1698 | Silent Mutation | TGC,TGT | C,C 587 | NP_003603.1 |