Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATGTGCCATCGCTGTGGTGAGG[C/T]ATGTGGTTGCTGTGGTGGGAAACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609291 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPRED1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPRED1 - sprouty related EVH1 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152594.2 | 1327 | Missense Mutation | GCA,GTA | A,V 432 | NP_689807.1 | |
XM_005254202.3 | 1327 | Missense Mutation | GCA,GTA | A,V 444 | XP_005254259.1 | |
XM_011521289.2 | 1327 | Missense Mutation | GCA,GTA | A,V 411 | XP_011519591.1 |