Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGTTGTCTGGGAGAGACAGGGA[C/T]CTGGAGGAAGGGCATACCTTTCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GRAMD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRAMD2 - GRAM domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012642.2 | 859 | Silent Mutation | AGA,AGG | R,R 217 | NP_001012660.1 | |
XM_011521327.2 | 859 | Silent Mutation | AGA,AGG | R,R 216 | XP_011519629.1 | |
XM_011521328.2 | 859 | Silent Mutation | AGA,AGG | R,R 215 | XP_011519630.1 | |
XM_011521329.2 | 859 | Silent Mutation | AGA,AGG | R,R 207 | XP_011519631.1 | |
XM_011521330.2 | 859 | Silent Mutation | AGA,AGG | R,R 205 | XP_011519632.1 | |
XM_011521331.2 | 859 | Silent Mutation | AGA,AGG | R,R 198 | XP_011519633.1 | |
XM_011521332.2 | 859 | Silent Mutation | AGA,AGG | R,R 181 | XP_011519634.1 | |
XM_011521333.2 | 859 | Silent Mutation | AGA,AGG | R,R 181 | XP_011519635.1 | |
XM_017021997.1 | 859 | Silent Mutation | AGA,AGG | R,R 179 | XP_016877486.1 | |
XM_017021998.1 | 859 | Silent Mutation | AGA,AGG | R,R 179 | XP_016877487.1 |