Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATCTCGAAGAACCAAGTCATTTA[A/C]AAGATATCTCTGCAGGGTCACGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118485 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC33 - coiled-coil domain containing 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CYP11A1 - cytochrome P450 family 11 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000781.2 | 1424 | Missense Mutation | GTA,TTA | V,L 399 | NP_000772.2 | |
NM_001099773.1 | 1424 | Missense Mutation | GTA,TTA | V,L 241 | NP_001093243.1 |