Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGGGTTACATGGAATCTACCTG[A/C]GGGCCTTCTGCACAGGGCTGGATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609610 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUBGCP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TUBGCP4 - tubulin gamma complex associated protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286414.2 | 555 | Silent Mutation | AGG,CGG | R,R 87 | NP_001273343.1 | |
NM_014444.4 | 555 | Silent Mutation | AGG,CGG | R,R 87 | NP_055259.2 | |
XM_011521454.2 | 555 | Silent Mutation | AGG,CGG | R,R 87 | XP_011519756.1 | |
XM_011521455.2 | 555 | UTR 5 | XP_011519757.1 | |||
XM_017022078.1 | 555 | Silent Mutation | AGG,CGG | R,R 87 | XP_016877567.1 |
ZSCAN29 - zinc finger and SCAN domain containing 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |