Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCCCCTCCATCAGGCCGCACCC[A/C]TGAAGTGGCAGCACGGAGAGCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604184 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGDCC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGDCC3 - immunoglobulin superfamily DCC subclass member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004884.3 | 2456 | Missense Mutation | NP_004875.2 | |||
XM_011522241.1 | 2456 | Missense Mutation | XP_011520543.1 | |||
XM_011522243.1 | 2456 | Missense Mutation | XP_011520545.1 | |||
XM_011522244.1 | 2456 | Missense Mutation | XP_011520546.1 | |||
XM_011522245.2 | 2456 | Intron | XP_011520547.1 |