Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAGATGAATCGGTTGGCATGGG[A/G]AGCTTTTAACTGGCCCCACTCCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611861 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADPGK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADPGK - ADP dependent glucokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_031284.4 | 764 | Missense Mutation | CCC,TCC | P,S 224 | NP_112574.3 | |
XM_006720712.3 | 764 | Missense Mutation | CCC,TCC | P,S 224 | XP_006720775.1 | |
XM_006720713.3 | 764 | Missense Mutation | CCC,TCC | P,S 102 | XP_006720776.1 | |
XM_011522102.2 | 764 | Intron | XP_011520404.1 | |||
XM_011522103.2 | 764 | Intron | XP_011520405.1 | |||
XM_011522104.2 | 764 | Missense Mutation | CCC,TCC | P,S 224 | XP_011520406.1 | |
XM_017022664.1 | 764 | Intron | XP_016878153.1 |