Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCACAAGCAGACGGAGAAGACGAC[C/G]CGCGTGCGGACTGTGCTGAACGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609481 MIM: 611611 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ISL2 - ISL LIM homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145805.2 | 736 | Silent Mutation | ACC,ACG | T,T 192 | NP_665804.1 | |
XM_017022505.1 | 736 | Silent Mutation | ACC,ACG | T,T 192 | XP_016877994.1 |
SCAPER - S-phase cyclin A associated protein in the ER | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |