Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCTCTGCCCCAGCGTCCACATA[A/G]CCAGCGGACAGCCTACGATGGAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612039 MIM: 608963 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LPCAT4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LPCAT4 - lysophosphatidylcholine acyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153613.2 | 1119 | Silent Mutation | GGC,GGT | G,G 341 | NP_705841.2 | |
XM_006720454.2 | 1119 | Silent Mutation | GGC,GGT | G,G 341 | XP_006720517.1 | |
XM_011521422.1 | 1119 | Silent Mutation | GGC,GGT | G,G 173 | XP_011519724.1 |
NUTM1 - NUT midline carcinoma family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |