Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTACTTCTCCCTGAGCCTCCTGC[A/G]GATCATGGACGTGGTGAGTCCCGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118511 | ||||||||||||||||||||
Literature Links: |
CHRNA7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRNA7 - cholinergic receptor nicotinic alpha 7 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000746.5 | 232 | Missense Mutation | CAG,CGG | Q,R 61 | NP_000737.1 | |
NM_001190455.2 | 232 | Missense Mutation | CAG,CGG | Q,R 90 | NP_001177384.1 | |
XM_011521176.2 | 232 | Intron | XP_011519478.2 | |||
XM_011521177.2 | 232 | Intron | XP_011519479.1 | |||
XM_011521178.2 | 232 | Missense Mutation | CAG,CGG | Q,R 61 | XP_011519480.1 | |
XM_017021882.1 | 232 | UTR 5 | XP_016877371.1 | |||
XM_017021883.1 | 232 | UTR 5 | XP_016877372.1 | |||
XM_017021884.1 | 232 | UTR 5 | XP_016877373.1 |
LOC105370940 - uncharacterized LOC105370940 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011522320.2 | 232 | Intron | XP_011520622.1 |