Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCATCTTCCTGAGATCTGAAGTC[C/T]GGAGACTAGCAGCCTGCTGCATGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612196 MIM: 180090 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABHD2 - abhydrolase domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RLBP1 - retinaldehyde binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000326.4 | 866 | Missense Mutation | CAG,CGG | Q,R 216 | NP_000317.1 | |
XM_011521870.2 | 866 | Missense Mutation | CAG,CGG | Q,R 216 | XP_011520172.1 | |
XM_017022460.1 | 866 | Missense Mutation | CAG,CGG | Q,R 225 | XP_016877949.1 |