Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCCAGCACACGTAGGCCCTGCAG[A/G]CTGCGGGCCCGAGAAAGGGCCACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610953 | ||||||||||||||||||||
Literature Links: |
PIF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PIF1 - PIF1 5'-to-3' DNA helicase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286496.1 | 1804 | Silent Mutation | NP_001273425.1 | |||
NM_001286497.1 | 1804 | Silent Mutation | NP_001273426.1 | |||
NM_001286499.1 | 1804 | Silent Mutation | NP_001273428.1 | |||
NM_025049.3 | 1804 | Silent Mutation | NP_079325.2 | |||
XM_011522083.2 | 1804 | Silent Mutation | XP_011520385.1 | |||
XM_011522084.2 | 1804 | Silent Mutation | XP_011520386.1 |