Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTTTAGGCCTGGGAATCAGGAG[C/T]GCTATGACAATTTCCTCCAAAGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600839 | ||||||||||||||||||||
Literature Links: |
CTXN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CTXN2 - cortexin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC12A1 - solute carrier family 12 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000338.2 | 421 | Missense Mutation | CGC,TGC | R,C 69 | NP_000329.2 | |
NM_001184832.1 | 421 | Missense Mutation | CGC,TGC | R,C 69 | NP_001171761.1 | |
XM_005254606.1 | 421 | Missense Mutation | CGC,TGC | R,C 69 | XP_005254663.1 |