Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCCTCCTTCTTGGCCTTCTTAGGA[A/G]TTCTTCTGGGGGATCTTTACTGTCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603687 | ||||||||||||||||||||
Literature Links: |
ALDH1A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH1A2 - aldehyde dehydrogenase 1 family member A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206897.1 | 1824 | Silent Mutation | AAC,AAT | N,N 496 | NP_001193826.1 | |
NM_003888.3 | 1824 | Silent Mutation | AAC,AAT | N,N 517 | NP_003879.2 | |
NM_170696.2 | 1824 | Silent Mutation | AAC,AAT | N,N 479 | NP_733797.1 | |
NM_170697.2 | 1824 | Silent Mutation | AAC,AAT | N,N 421 | NP_733798.1 |