Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGTACGGAGACCCCATGTGCAC[A/G]GGCGACTTCATTTCTACATAGCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612454 MIM: 605570 | ||||||||||||||||||||
Literature Links: |
MEGF11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MEGF11 - multiple EGF like domains 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032445.2 | 3474 | Silent Mutation | CCC,CCT | P,P 968 | NP_115821.2 | |
XM_017022670.1 | 3474 | Silent Mutation | CCC,CCT | P,P 1021 | XP_016878159.1 | |
XM_017022671.1 | 3474 | Silent Mutation | CCC,CCT | P,P 1064 | XP_016878160.1 | |
XM_017022672.1 | 3474 | Silent Mutation | CCC,CCT | P,P 968 | XP_016878161.1 | |
XM_017022673.1 | 3474 | Intron | XP_016878162.1 | |||
XM_017022674.1 | 3474 | Silent Mutation | CCC,CCT | P,P 778 | XP_016878163.1 | |
XM_017022675.1 | 3474 | Silent Mutation | CCC,CCT | P,P 778 | XP_016878164.1 |
RAB11A - RAB11A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |