Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGTATGTCTCTGAGACTGTGGAC[A/G]TGCCATCCCCATTTGACCTACTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600178 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAP1A - microtubule associated protein 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002373.5 | 806 | Missense Mutation | ATG,GTG | M,V 17 | NP_002364.5 | |
XM_017022189.1 | 806 | Missense Mutation | ATG,GTG | M,V 255 | XP_016877678.1 |