Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGCCCAGGGCCACCCACACGCA[A/C]GGCGCCCTGATGGAGACGCTGCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613298 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TICRR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TICRR - TOPBP1 interacting checkpoint and replication regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308025.1 | 303 | Missense Mutation | CAA,CAC | Q,H 101 | NP_001294954.1 | |
NM_152259.3 | 303 | Missense Mutation | CAA,CAC | Q,H 101 | NP_689472.3 |