Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGCTGGTGGACTTGAAGATCAT[C/T]TGGAATAAGACCAAGCATGACGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612799 | ||||||||||||||||||||
Literature Links: |
EARS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EARS2 - glutamyl-tRNA synthetase 2, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083614.1 | 481 | Intron | NP_001077083.1 | |||
NM_001308211.1 | 481 | Intron | NP_001295140.1 | |||
XM_011545738.1 | 481 | Intron | XP_011544040.1 | |||
XM_011545739.1 | 481 | Intron | XP_011544041.1 |
UBFD1 - ubiquitin family domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019116.2 | 481 | Silent Mutation | ATC,ATT | I,I 91 | NP_061989.2 | |
XM_011545894.2 | 481 | Silent Mutation | ATC,ATT | I,I 91 | XP_011544196.1 | |
XM_017023459.1 | 481 | Silent Mutation | ATC,ATT | I,I 91 | XP_016878948.1 | |
XM_017023460.1 | 481 | Silent Mutation | ATC,ATT | I,I 91 | XP_016878949.1 |