Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGGGACACCAGTGAGGATGCCT[C/T]GATCCATCGATTGGAAGGCACTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605584 | ||||||||||||||||||||
Literature Links: |
DHX38 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHX38 - DEAH-box helicase 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014003.3 | 308 | Missense Mutation | TCG,TTG | S,L 9 | NP_054722.2 | |
XM_005256269.1 | 308 | Missense Mutation | TCG,TTG | S,L 9 | XP_005256326.1 | |
XM_011523484.1 | 308 | Missense Mutation | TCG,TTG | S,L 9 | XP_011521786.1 | |
XM_011523485.1 | 308 | Missense Mutation | TCG,TTG | S,L 9 | XP_011521787.1 | |
XM_017023913.1 | 308 | Missense Mutation | TCG,TTG | S,L 9 | XP_016879402.1 |
TXNL4B - thioredoxin like 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |