Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGAGTCATAAGGAGGAGCCGTGG[A/G]GTCATTGTCTGCCTCCTGTATTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600023 | ||||||||||||||||||||
Literature Links: |
CDH11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDH11 - cadherin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308392.1 | 2749 | UTR 3 | NP_001295321.1 | |||
NM_001797.3 | 2749 | Missense Mutation | CCC,TCC | P,S 732 | NP_001788.2 | |
XM_005255761.3 | 2749 | Missense Mutation | CCC,TCC | P,S 732 | XP_005255818.1 | |
XM_005255762.2 | 2749 | Missense Mutation | CCC,TCC | P,S 732 | XP_005255819.1 | |
XM_005255763.2 | 2749 | Missense Mutation | CCC,TCC | P,S 732 | XP_005255820.1 | |
XM_011522803.2 | 2749 | Missense Mutation | CCC,TCC | P,S 732 | XP_011521105.1 | |
XM_017022830.1 | 2749 | Missense Mutation | CCC,TCC | P,S 606 | XP_016878319.1 |