Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAGACTGAAGAACAGATGGAGGG[A/G]GGCTGGTGTTGGCAAGCAGCCGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611675 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM92B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM92B - family with sequence similarity 92 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198491.2 | 875 | Missense Mutation | CCC,CTC | P,L 240 | NP_940893.1 | |
XM_005255931.2 | 875 | Missense Mutation | CCC,CTC | P,L 240 | XP_005255988.1 | |
XM_011523063.1 | 875 | Missense Mutation | CCC,CTC | P,L 240 | XP_011521365.1 | |
XM_017023198.1 | 875 | Missense Mutation | CCC,CTC | P,L 240 | XP_016878687.1 |
KIAA0513 - KIAA0513 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |