Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCCTCACCATGTTCATCCTGGAC[A/G]ACCATGATTCAGACCTAGACCTTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC400541 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC400541 - uncharacterized LOC400541 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDPR - pyruvate dehydrogenase phosphatase regulatory subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322117.1 | 2927 | Missense Mutation | AAC,GAC | N,D 774 | NP_001309046.1 | |
NM_001322118.1 | 2927 | Missense Mutation | AAC,GAC | N,D 674 | NP_001309047.1 | |
NM_001322119.1 | 2927 | Missense Mutation | AAC,GAC | N,D 372 | NP_001309048.1 | |
NM_017990.4 | 2927 | Missense Mutation | AAC,GAC | N,D 774 | NP_060460.4 | |
XM_005256015.2 | 2927 | Missense Mutation | AAC,GAC | N,D 774 | XP_005256072.1 | |
XM_005256016.3 | 2927 | Missense Mutation | AAC,GAC | N,D 774 | XP_005256073.1 | |
XM_006721224.2 | 2927 | Missense Mutation | AAC,GAC | N,D 713 | XP_006721287.1 | |
XM_006721225.3 | 2927 | Intron | XP_006721288.1 | |||
XM_011523190.2 | 2927 | Intron | XP_011521492.1 | |||
XM_017023387.1 | 2927 | Missense Mutation | AAC,GAC | N,D 774 | XP_016878876.1 | |
XM_017023388.1 | 2927 | Intron | XP_016878877.1 | |||
XM_017023389.1 | 2927 | Intron | XP_016878878.1 |