Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGGGCTCCGAGACAGACCTATA[G/T]GCTGAGGCTGGTACCCCCTCTGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603464 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK10 - cyclin dependent kinase 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPATA2L - spermatogenesis associated 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152339.3 | 1539 | Silent Mutation | NP_689552.2 | |||
XM_005256279.4 | 1539 | Silent Mutation | XP_005256336.1 |
VPS9D1 - VPS9 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |