Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGATGTAGAGGTCTCCGTGCTCGC[A/C]GTAGATGGCGTTGTCCCGGTCGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614147 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1QTNF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1QTNF8 - C1q and tumor necrosis factor related protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207419.3 | 968 | Missense Mutation | AGC,CGC | S,R 232 | NP_997302.2 | |
XM_011522483.2 | 968 | Missense Mutation | AGC,CGC | S,R 232 | XP_011520785.1 |