Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTGAAACAGAAATTCTGGAATA[C/G]CTATCTGGTAAGATAGGCGTTTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MPV17L PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPV17L - MPV17 mitochondrial inner membrane protein like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128423.1 | 518 | Missense Mutation | ACC,AGC | T,S 125 | NP_001121895.1 | |
NM_173803.3 | 518 | Intron | NP_776164.2 | |||
XM_017023108.1 | 518 | Intron | XP_016878597.1 |