Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAGCCTGCTGGTGCTGCGCCGC[C/T]GGCCCTGCTCCCGCGACGGCTCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608508 MIM: 604628 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYBA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYBA - cytochrome b-245 alpha chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL17C - interleukin 17C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013278.3 | 530 | Missense Mutation | CGG,TGG | R,W 161 | NP_037410.1 |
ZC3H18 - zinc finger CCCH-type containing 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |