Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAGCACAGAACCTGGTGCTGAT[C/T]CCGCTGCACGCGGCGCCGCATCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602622 MIM: 603022 MIM: 600305 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNASE1L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNASE1L2 - deoxyribonuclease I-like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301680.1 | 611 | Silent Mutation | ATC,ATT | I,I 167 | NP_001288609.1 | |
NM_001374.2 | 611 | Silent Mutation | ATC,ATT | I,I 167 | NP_001365.1 | |
XM_011522399.1 | 611 | Silent Mutation | ATC,ATT | I,I 258 | XP_011520701.1 |
E4F1 - E4F transcription factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ECI1 - enoyl-CoA delta isomerase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |