Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAATGGCTGAGCCTTATAACCTA[G/T]CTCTTTCTTGGCTTTCTCTAAGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616164 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SDR42E1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SDR42E1 - short chain dehydrogenase/reductase family 42E, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145168.2 | 1131 | Missense Mutation | NP_660151.2 | |||
XM_005256257.3 | 1131 | Missense Mutation | XP_005256314.1 | |||
XM_011523471.2 | 1131 | Missense Mutation | XP_011521773.1 | |||
XM_011523472.2 | 1131 | Missense Mutation | XP_011521774.1 |