Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGGACGTGCGGGCCTTGGGCAC[C/G]GAACCCCACTTCTTTGACAGGAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604056 | ||||||||||||||||||||
Literature Links: |
HS3ST2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HS3ST2 - heparan sulfate-glucosamine 3-sulfotransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006043.1 | 884 | Silent Mutation | ACC,ACG | T,T 145 | NP_006034.1 | |
XM_011546001.2 | 884 | Silent Mutation | ACC,ACG | T,T 145 | XP_011544303.1 | |
XM_011546002.2 | 884 | Silent Mutation | ACC,ACG | T,T 145 | XP_011544304.1 |
LOC107984851 - collagen alpha-1(XVI) chain-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017023983.1 | 884 | Intron | XP_016879472.1 |