Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGCGTCACCTCCTTCTGCCCCACC[C/T]TGGTCACTTCCCCACCGGAGGTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 108745 MIM: 611113 | ||||||||||||||||||||
Literature Links: |
AMDHD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMDHD2 - amidohydrolase domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145815.1 | 425 | Silent Mutation | CTG,TTG | L,L 110 | NP_001139287.1 | |
NM_015944.3 | 425 | Silent Mutation | CTG,TTG | L,L 110 | NP_057028.2 | |
XM_017023262.1 | 425 | Silent Mutation | CTG,TTG | L,L 110 | XP_016878751.1 | |
XM_017023263.1 | 425 | Silent Mutation | CTG,TTG | L,L 110 | XP_016878752.1 | |
XM_017023264.1 | 425 | Silent Mutation | CTG,TTG | L,L 110 | XP_016878753.1 | |
XM_017023265.1 | 425 | Silent Mutation | CTG,TTG | L,L 110 | XP_016878754.1 | |
XM_017023266.1 | 425 | UTR 5 | XP_016878755.1 | |||
XM_017023267.1 | 425 | UTR 5 | XP_016878756.1 |
ATP6V0C - ATPase H+ transporting V0 subunit c | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198569.1 | 425 | Intron | NP_001185498.1 | |||
NM_001694.3 | 425 | Intron | NP_001685.1 |
CEMP1 - cementum protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |