Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGGGTGTCATCAATGCAGAGAT[A/C]CCCAACTGCTGGGAGTGCCCTCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609085 | ||||||||||||||||||||
Literature Links: |
FBXL19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXL19 - F-box and leucine rich repeat protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099784.2 | 441 | Silent Mutation | ATA,ATC | I,I 139 | NP_001093254.2 | |
NM_001282351.1 | 441 | UTR 5 | NP_001269280.1 |
FBXL19-AS1 - FBXL19 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |