Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCTCACAAGGGAGGAGGGTGCC[A/G]AGGGCCGGCCATGGGCCCTTTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603213 MIM: 600999 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIF22 - kinesin family member 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256269.1 | 674 | Missense Mutation | AAG,GAG | K,E 96 | NP_001243198.1 | |
NM_001256270.1 | 674 | Missense Mutation | AAG,GAG | K,E 96 | NP_001243199.1 | |
NM_007317.2 | 674 | Missense Mutation | AAG,GAG | K,E 164 | NP_015556.1 |
MAZ - MYC associated zinc finger protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |