Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGATCTTACCCAAAGACTTGAACA[C/T]GGTGGTCTTCTCACAGTGGGCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609901 MIM: 123740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKS4B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKS4B - ankyrin repeat and sterile alpha motif domain containing 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CRYM - crystallin mu | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001888.4 | 1216 | Missense Mutation | ATG,GTG | M,V 289 | NP_001879.1 |